Documenti fondamentali
Z684554
Ace Rotary Evaporator Vial Adapter Kit
FETFE® O-ring, adapter joint: ST/NS 29/32
Sinonimo/i:
Adapter Kit for Vials, Rotary Evaporator Adapter Kit, Vial Adapter Kit
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Materiali
FETFE® O-ring
Giunto
joint: ST/NS adapter29/32
Descrizione
PTFE adapters with #15 Ace-Thred to GPI vial thread, sizes 8-425, 13-425, 15-425, 18-400, 20-400, 22-400, 24-410
Produttore/marchio commerciale
Ace Glass 13290-65
Categorie correlate
Descrizione generale
PTFE connecting adapters and glass conversion adapters for connecting GPI threaded vials to a rotary evaporator vapor tube or trap. Both are suitable for vacuum work. PTFE connecting adapters are available with FETFE® or Chemraz® O-rings. Order Chemraz if you are using methylene chloride or acetone.
Note legali
Ace-Thred is a trademark of Ace Glass, Inc.
Chemraz is a registered trademark of Greene, Tweed of Delaware, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
Z684554-1EA4061832353203