Documenti fondamentali
Y907227
Sigma-Aldrich
Valve
brass diaphragm, for CGA 350 right angle path, external, left hand thread
About This Item
Codice UNSPSC:
12142200
NACRES:
NA.12
Materiali
brass diaphragm
Livello qualitativo
Parametri
2640 psi max. pressure
Lunghezza
4.62 in.
Peso
613 g
Raccordi
for CGA 350 right angle path, external, left hand thread
Categorie correlate
Altre note
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)