WHA10461300
Whatman® FP 050/0 filter holder
FP050/0, upper section only
Sinonimo/i:
Z698520
About This Item
Codice UNSPSC:
41104922
NACRES:
NB.24
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Whatman 10461300
Whatman Article No. 28415726 (US reference)
Categorie correlate
Descrizione generale
The female Luer lock inlet is compatible with typical syringes. Polysulfone housing can be autoclaved with membrane in place for applications requiring a sterile filter assembly.
Caratteristiche e vantaggi
- Durable polysulfone plastic construction provides moderate chemical compatibility with lighter weight than stainless steel filter holders.
- Polysulfone filter holders are often used for clarification, venting applications, particulate removal or collection from aqueous, air or gas streams.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)