Documenti fondamentali
WHA10461200
Whatman® FP 050/0 filter holder
polysulfone, for 50 mm filters
Sinonimo/i:
Z751456
About This Item
Codice UNSPSC:
41104922
NACRES:
NB.24
Confezionamento
pk of 5
Produttore/marchio commerciale
Whatman 10461200
Whatman Article No. 28415725 (US reference)
Descrizione generale
The female Luer lock inlet is compatible with typical syringes. Polysulfone housing can be autoclaved with membrane in place for applications requiring a sterile filter assembly.
Caratteristiche e vantaggi
  • Durable polysulfone plastic construction provides moderate chemical compatibility with lighter weight than stainless steel filter holders.
  • Polysulfone filter holders are often used for clarification, venting applications, particulate removal or collection from aqueous, air or gas streams.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2