Documenti fondamentali
WHA10442101
Whatman® glass vacuum filtration device
GV050 series, GV050/1/03 support screen
Sinonimo/i:
Z750859
About This Item
Codice UNSPSC:
41104900
NACRES:
NB.24
Materiali
borosilicate glass funnel(s) (funnel)
stainless steel clamp(s) (clamp)
Confezionamento
pk of 1
Produttore/marchio commerciale
Whatman 10442101
Whatman Article No. 28415562 (US reference)
Compatibilità
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
Descrizione generale
Whatman® Vacuum Type Glass Holders
Produced from borosilicate glass and available with a choice of support screen. Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
The sintered glass support is recommended for filtration and biological analysis. The 304 stainless steel support screen is suitable for use with proteinaceous solutions.
Caratteristiche e vantaggi
  • Accessory to GV050 Glass Filter Funnel
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)