WAMUPG2HCPWC1
Whatman® Mini-UniPrep® G2 hand compressor
1 position, pack of 1
Sinonimo/i:
hand compressor, mini-uniprep G2, syringeless filter, whatman filter
About This Item
Codice UNSPSC:
40161807
NACRES:
NB.24
Prodotti consigliati
Confezionamento
pack of 1
Produttore/marchio commerciale
Whatman Article No. 29026195 (US reference)
Whatman MUPG2HCPWC1
Descrizione generale
Mini-UniPrep G2 Hand Compressor is for use with Mini-UniPrep G2 Syringeless filters.
Caratteristiche e vantaggi
- Ergonomic hand compressor allows for sample preparation with increased hand comfort
- Mini-UniPrepAll-in-One syringeless filter prepares samples three times faster than standard syringe filter method
- Mini-UniPrepglass or polypropylene vials support analyses such as pharmaceutical quality control (pharma QC) and water monitoring
- Mini-UniPrepvials can be loaded directly into the autosampler
Note legali
Mini-UniPrep is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)