WHAMUPG2MCWT8
Whatman® Mini-UniPrep® G2 hand compressor
8 position tray, pack of 1
Sinonimo/i:
hand compressor, mini-uniprep G2, syringeless filter, whatman filter
About This Item
Codice UNSPSC:
40161807
NACRES:
NB.24
Prodotti consigliati
Confezionamento
pack of 1
Produttore/marchio commerciale
Cytiva
Whatman Article No. 29036891 (US reference)
Whatman MUPG2MCWT8
Categorie correlate
Descrizione generale
Mini-UniPrep G2 Hand Compressor is for use with Mini-UniPrep G2 Syringeless filters.
Caratteristiche e vantaggi
- A multicompressor streamlines syringeless filtration – preload in advance with an extra tray
- Designed for Cytiva′s Mini-UniPrepG2, which combines a borosilicate glass vial, plunger and attached filter membrane, and septum with cap
Note legali
Mini-UniPrep is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)