TEM-7435703
Carbon Film Supported Copper Tomography Grid R14C2
pack of 25 ×
Sinonimo/i:
Carbon Film on Tomography Grid, 25pk
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Prodotti consigliati
Confezionamento
pack of 25 ×
Descrizione generale
This 1.5mm square, 300 mesh grid is made from copper, supported by a carbon film, and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90°.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
TEM-7435703 | 4061841757993 |