Documenti fondamentali
SYNF68272L
Synthware Sure-Grip separatory funnel
2000 mL, stopper size #27
Sinonimo/i:
Synthware F68272L
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.51
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F68272L
Capacità
2000 mL
2000 mL
Descrizione generale
Globe shaped separatory funnel with a long stopper neck and straight section between the funnel body and stopcock, which provides a secure grip while shaking and also aids in clamping. Available with PTFE stopcocks. Supplied complete with one listed size PTFE or glass stopper.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)