Documenti fondamentali
SYNF606150C
Synthware Buchner funnel
150 mL, porosity: coarse
Sinonimo/i:
Synthware F606150C, filtering funnel
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.51
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F606150C
Capacità
150 mL
150 mL
Descrizione generale
With sealed-in glass disc of the listed porosity. Not recommended for filtration where pressure would exceed 15psi. XC-extra coarse, C-Coarse, M-Medium, F-Fine porosity.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)