Documenti fondamentali
SYNF490250
Synthware storage flask with high vacuum valve and side arm
250 mL, valve size 0-4 mm
Sinonimo/i:
Synthware F490250, round bottom flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.51
Materiali
borosilicate glass flask
round bottom
single-neck (high vacuum valve)
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F490250
Capacità
250 mL
250 mL
Misura valvola
0-4 mm
Descrizione generale
Supplied with high vacuum valve with PTFE O-ring at tip and PTFE wiper( “umbrella”) to protect Viton® O-ring on shaft..
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)