Key Documents
SYNC579110
Synthware water jacketed distillation column with removable hose connections
joint: ST/NS 19/22, jacket H 110 mm
Sinonimo/i:
Synthware C579110, glass distilling column
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Giunto
joint: ST/NS19/22
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware C579110
Altezza camicia
110 mm
Descrizione generale
Indentations at the bottom for supporting packing material. Jacket permits the column to be used as a condenser.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)