Key Documents
SYNA622440
Synthware vacuum take-off adapter with short stem
joint: ST/NS 24/40, hose size 10 mm
Sinonimo/i:
Synthware A622440, distillation adapter, vacuum take off adapter
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A622440
Dimens. tubo
10 mm
Descrizione generale
With a hose connection and a drip tip.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)