SYNA751922
Synthware™ 90 degree vacuum adapter with glass stopcock
male joint: ST/NS 19/22
Sinonimo/i:
Synthware A751922, flow control adapter
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Prodotti consigliati
Giunto
male joint: ST/NS19/22
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A751922
Categorie correlate
Descrizione generale
Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2 mm.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)