Documenti fondamentali
SYNA381410
Synthware vacuum adapter with PTFE stopcock
joint: ST/NS 14/10
Sinonimo/i:
Synthware A381410, glass adapter with stopcock
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Giunto
joint: ST/NS14/10
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
A381410
Descrizione generale
Synthware vacuum adapter with PTFE stopcock for microscale applications. Hose connection is 8 mm O.D. and joint is 14/10. Stopcock bore is 2 mm.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)