SLW1003-5L
Pyrex® Griffin beakers, heavy duty, graduated
5000 mL
Sinonimo/i:
Pyrex, SciLabware 1003-5L
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
white graduation
Caratteristiche
graduated
heavy duty
Produttore/marchio commerciale
1003-5L
SciLabware
Alt. × Diam.
300 mm × 180 mm
Capacità
5000 mL
Descrizione generale
- Mechanically stronger than standard beakers
- Designed for longer life and increased user safety
- Reinforced rims and spout, less prone to breakage during washing etc.
- Thicker walls and heavier bases
- Manufactured from Pyrex borosilicate glass
- White graduations and marking spot
Altre note
Browse all glass beakers
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SLW1003-5L-1EA | 4061832435626 |