BR91273
BRAND® glass beaker with spout, low form
volume 5000 mL, with graduation
Sinonimo/i:
Beakers, Glass
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass 3.3
Caratteristiche
with graduation
Produttore/marchio commerciale
BRAND 91273
Volume
5000 mL
Categorie correlate
Descrizione generale
The BRAND® Glass Beaker with Spout is expertly crafted from Boro 3.3 glass, making it a reliable choice for laboratory applications. Designed in accordance with DIN EN ISO 3819 standards, this low-form beaker features clear graduation markings for precise measurement of liquids. The integrated spout allows for easy pouring, minimizing spills and enhancing accuracy during liquid transfer.
Altre note
Browse all glass beakers
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
BR91273-1EA | 4061831861709 |