Documenti fondamentali
MT30019823
Mettler-Toledo SevenCompact uPlace electrode holder, (included in standard delivery)
Sinonimo/i:
electrode stand
About This Item
Codice UNSPSC:
41115603
NACRES:
NB.79
Produttore/marchio commerciale
Mettler-Toledo 30019823
Descrizione generale
  • Mettler-Toledo SevenCompact uPlace electrode holder is an accessory for use with the pH meter.
  • The electrode arm of the holder can be moved straight up and down and can be operated with one hand.
  • Works more efficiently when compared to the traditional designs.
Note legali
SevenCompact is a trademark of Mettler-Toledo, Inc.
uPlace is a trademark of Mettler-Toledo, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
MT30019823-1EA