DWK2515415-B
Kimble® Closed Top Linerless Polypropylene Screw Thread Cap
15-415, black screw cap
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.17
Prodotti consigliati
Materiali
black screw cap
red polypropylene screw cap (Push fit)
Confezionamento
pkg of 1 packs
Produttore/marchio commerciale
Kimble® 2515415-B
Raccordi
thread
Categorie correlate
Applicazioni
Closure solution for vials and bottles
Caratteristiche e vantaggi
- Economical, one-piece construction
- Unique design provides exceptional sealing properties
- Autoclavable
- Designed from ASTM Specification E982
Note legali
KIMBLE is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)