Documenti fondamentali
DWK73808-15415
Kimble® Closed Top Polypropylene Screw Thread Cap With Welded Ptfe-Faced Silicone Liner
15-415
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.17
Materiali
PTFE/silicone liner
red polypropylene screw cap (Push fit)
Confezionamento
pkg of 1 packs
Produttore/marchio commerciale
Kimble® 73808-15415
Raccordi
thread
Applicazioni
Closure solution for vials and bottles
Caratteristiche e vantaggi
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Closures are ideal for repeated autoclaving
  • Welded liner technology eliminates the possibility of glue contamination
Note legali
KIMBLE is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)