Documenti fondamentali
DWK11-0054G
Wheaton® Microliter Snap Caps With Septa
green PTFE/silicone cap
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.11
Materiali
green PTFE/silicone cap
Caratteristiche
cap pre-cut
Confezionamento
pkg of 100 ea
Produttore/marchio commerciale
WHEATON® 11-0054G
Applicazioni
Chromatography
Caratteristiche e vantaggi
  • 11mm Snap Caps with installed septa offered in a wide variety of the most popular configurations.
  • Where noted, the Ring is installed between cap and septa for robotics employing magnetics to move vials.
  • Approved for GC and LC (where noted).
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)