HS234634
Pipette Tip Disposal Box
clear (disposal box)
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.01
Prodotti consigliati
Materiali
acrylic
clear (disposal box)
Caratteristiche
Pipette tips
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Heathrow Scientific HS234634
Lungh. × Largh. × Alt.
154 mm × 171 mm × 206 mm
Descrizione generale
- The 3/8 in. (10 mm) thick acrylic provides protection from Beta radiation
- The large, hinged lid allows easy disposal of contents
- Use in conjunction with small biohazard disposal bags, if hazardous fluids are not acrylic friendly
The Pipette Tip Disposal Box′s quick flip of port cover makes tip ejection easy. It is convenient for one-handed disposal of used pipette tips and eliminates any need to handle pipette tips used with hazardous. fluids.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)