Documenti fondamentali
CLS361125
Pyrex® moisture test distilling receiver, 24/40 joints
capacity 25 mL
Sinonimo/i:
Corning® 3611
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Produttore/marchio commerciale
Corning 3611-25
Altezza
317 mm
Capacità
25 mL
Descrizione generale
Graduated, from 0 to 1 mL, in 0.1 mL intervals, with a maximum scale error of 0.05 mL. Graduation intervals from 1.0 to 10.0 mL are 0.2 mL, with a maximum scale error of 0.1 mL.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)