CLS31668
Pyrex® cloning cylinder
O.D. × H 8 mm × 8 mm
Sinonimo/i:
lab equipment accessory
About This Item
Codice UNSPSC:
41106618
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
cylindrical
Sterilità
non-sterile
Confezionamento
pack of 125
Produttore/marchio commerciale
Corning 3166-8
D.E. × alt.
8 mm × 8 mm
Descrizione generale
Clone a single cell or group of cells by surrounding them with this Pyrex glass open ended cloning cylinder. Dip the end of the cylinder into sterile silicone grease before pressing to the bottom of a culture dish to create an isolated well. Cylinders are autoclavable or can be dry heat sterilized.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Linlin Zhang et al.
PloS one, 11(5), e0154238-e0154238 (2016-05-18)
Porcine reproductive and respiratory syndrome virus (PRRSV) is one of the most devastating pathogens in the swine industry worldwide. Due to the lack of robust cell lines and small animal models, the pathogenesis of PRRSV infection and mechanism for protective
Sally Yunsun Kim et al.
Biomaterials, 267, 120480-120480 (2020-11-07)
Research into mechanisms underlying lung injury and subsequent repair responses is currently of paramount importance. There is a paucity of models that bridge the gap between in vitro and in vivo research. Such intermediate models are critical for researchers to
Global Trade Item Number
SKU | GTIN |
---|---|
CLS31668-125EA | 4061833883570 |