BR723016
BRAND® Haemacytometer cover glass, for counting chambers
borosilicate glass, size 22 mm × 30 mm × 0.4 mm
Sinonimo/i:
Cover glasses, Microscopy
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.22
Prodotti consigliati
Materiali
borosilicate glass
Caratteristiche
IVD
Produttore/marchio commerciale
BRAND 723016
Misura
22 mm × 30 mm × 0.4 mm
Tipo di binding
non-treated surface
Descrizione generale
Borosilicate glass, DIN ISO 8255. Refractive index ne = 1.52 ± 0.01, Abbe number ve = 56.5 ± 0.5. Flatness tolerance ± 3 μm.
They differ from ordinary cover glasses by their plain ground and polished surface.
They differ from ordinary cover glasses by their plain ground and polished surface.
Confezionamento
2 cover glasses in a plastic bag, separated by tissue paper.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)