BR721305
BRAND® counting chamber BLAUBRAND® Nageotte pattern
without clips, double ruled
Sinonimo/i:
BLAUBRAND®, Cell culture, Counting Chambers, Nageotte
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.22
Prodotti consigliati
Materiali
colorless
glass
Sterilità
non-sterile
Caratteristiche
without clips
Produttore/marchio commerciale
BRAND 721305
Pozzetti
1
Descrizione generale
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.The square area of 100 mm² is subdivided into 40 rectangles with an area of 2.5 mm² each.This chamber is commonly used for counting cells in the cerebro-spinal fluid, or for counting nematodes.
Caratteristiche e vantaggi
- BLAUBRAND® quality
- Outstanding measurement precision
- 100% certified quality
Confezionamento
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)