Documenti fondamentali
BR719005
BRAND® counting chamber BLAUBRAND® Malassez pattern
without clips, double ruled
Sinonimo/i:
BLAUBRAND®, Cell culture, Counting Chambers, Malassez
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.22
Materiali
colorless
glass
Sterilità
non-sterile
Caratteristiche
without clips
Produttore/marchio commerciale
BRAND 719005
Pozzetti
1
Descrizione generale
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.The ruling is rectangular, covering an area of 5 mm². The large rectangles measure 0.25 x 0.20 = 0.05 mm². They are each subdivided into 20 mini squares with an area of 0.0025 mm².This chamber is commonly used for counting cells in the cerebro-spinal fluid, or for counting nematodes.
Caratteristiche e vantaggi
  • BLAUBRAND® quality
  • Outstanding measurement precision
  • 100% certified quality
Confezionamento
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)