Documenti fondamentali
BR4600351
BRAND® Dispensette® S Digital bottle-top dispenser
volume range 2.5-25 mL, with recirculation valve
Sinonimo/i:
bottle top dispensers
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Caratteristiche
incremental 0.1 mL
DE-M marking
with recirculation valve
Produttore/marchio commerciale
BRAND 4600351
Intervallo di volume
2.5-25 mL
Raccordi
(GL 32-33)
GL 38
S 40
Descrizione generale
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Caratteristiche e vantaggi
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Altre note
Digital-The mechanical volume setting is easy to read and ensures accurate and reproducible volume control. Easy Calibration Technique for calibration adjustment in seconds without tools. A calibration flag indicates changes from factory settings.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Lot/Batch Number
Ci dispiace, ma al momento non ci sono COA disponibili online per questo prodotto.
Visita l’Archivio dei documenti
Protocolli
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.