BR362138
BRAND® BLAUBRAND® ASTM 100 mL centrifuge tube
pear-shaped, cylindrical, former standard ASTM D96
Sinonimo/i:
ASTM, BLAUBRAND®, Centrifuge Tubes, Tubes
About This Item
Codice UNSPSC:
41121703
NACRES:
NB.11
Prodotti consigliati
Materiali
borosilicate glass
cylindrical
pear-shaped
Sterilità
non-sterile
Caratteristiche
RCF 700 × g
Confezionamento
pack of 2
Produttore/marchio commerciale
BRAND 362138
Lunghezza
160 mm
Categorie correlate
Descrizione generale
Withstands RCF up to 700. Design, accuracy, etc. to meet ASTM ("American Society for Testing and Materials") specifications.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR362138-2EA | 4061831855814 |