Documenti fondamentali
BR27005
BRAND® SILBERBRAND ETERNA Graduated pipette, Class B, Type 2
2 mL with 0.01 mL graduations, not suitable for cotton plugs
Sinonimo/i:
ETERNA, Graduated Pipettes, Pipettes, SILBERBRAND
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Materiali
AR-Glass
Confezionamento
pack of 12 ea
Produttore/marchio commerciale
BRAND 27005
Campo di misura
accuracy: 0.01 mL
Capacità
2.0 mL
Descrizione generale
The BRAND® SILBERBRAND ETERNA graduated pipette is a Class B, Type 2 instrument made from AR-Glas® and complies with DIN EN ISO 835 standards. Designed for precision, it is calibrated to deliver (TD, Ex), ensuring accurate liquid transfer for various laboratory applications. This pipette combines durability with functionality, making it a reliable choice for professionals requiring high-quality measurement tools. Its clear graduation markings facilitate easy reading, enhancing user efficiency.
Note legali
AR-Glas is a registered trademark of Schott AG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
BR27005-12EA4061831845907