Documenti fondamentali
BR23830
BRAND® compact automatic SILBERBRAND burette, without support/clamp
volume 50 mL, with LDPE bottle and PVC filling tube
Sinonimo/i:
Automatic Burettes, Burettes, Compact Automatic Burettes, SILBERBRAND
About This Item
Codice UNSPSC:
41121808
NACRES:
NB.43
Materiali
borosilicate glass (5.4)
Produttore/marchio commerciale
BRAND 23830
Campo di misura del volume
50 mL, accuracy: ±0.10 mL
Volume
50 mL
Descrizione generale
The Modular Automatic Burette Concept! Fast to dismantle and easy to clean. Easy to repair - all individual components replaceable! Borosilicate glass 5.4, detachable PTFE stopcock. Calibrated to deliver (TD, Ex). 25 ml: Error limit according to class B, DIN EN ISO 385. 50 ml: DIN EN ISO 385, class B.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)