Documenti fondamentali
BR23810
BRAND® compact automatic SILBERBRAND burette, without support/clamp
volume 25 mL, with LDPE bottle and PVC filling tube
Sinonimo/i:
Automatic Burettes, Burettes, Compact Automatic Burettes, SILBERBRAND
About This Item
Codice UNSPSC:
41121808
NACRES:
NB.43
Materiali
AR-Glass
Produttore/marchio commerciale
BRAND 23810
Campo di misura del volume
25 mL, accuracy: ±0.08 mL
Volume
25 mL
Descrizione generale
The Modular Automatic Burette Concept! Fast to dismantle and easy to clean. Easy to repair - all individual components replaceable! Boro 3.3, detachable PTFE stopcock. Error limits according to class B, DIN EN ISO 385. Calibrated to deliver (TD, Ex).
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
BR23810-1EA4061831864809