Documenti fondamentali
BAH500250410
SP Bel-Art® Glove Box Sleeved Gloves for 8 in. Glove Ports
size 10 large
Sinonimo/i:
glove box
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.43
Materiali
black neoprene
Misura
10 large
Confezionamento
pkg of 1 pair
Produttore/marchio commerciale
Bel-Art H50025-0410
Descrizione generale
One-piece, seamless, smooth neoprene protection against liquid, gas or solid substance penetration.
Caratteristiche e vantaggi
  • Suitable for use with glove boxes having 8" glove ports including sid ENTRY H50026-0000 Glove Boxes
  • Requires clamping rings (H50025-0320) to hold gloves in place
  • Sold in pairs
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)