Z260517
sidEntry™ glove box
a clear acrylic glove box which features a large side entry, gasketed door, and a shelf rack
Sinonimo/i:
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
About This Item
Codice UNSPSC:
41103406
NACRES:
NB.35
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art H50026-0000
Lungh. × Largh. × Alt.
30 in. × 24 in. × 24 in.
Categorie correlate
Descrizione generale
The Scienceware sidEntry glove box is a clear acrylic glove box which features a large side entry, gasketed door, and a shelf rack to store small items. Glove box stability is assured by four adjustable levelers that also level the box and all its contents. The sidEntry glove box provides an isolated work environment for processing samples or handling air-sensitive materials while maintaining an anaerobic or other gas specific environment within the glove box.
- 8 in. glove ports accept standard dry-box gloves without clamps or O-rings
- Small enough to move from area to area
- Side door: 17 1/2 in. × 17 1/2 in.
Note legali
sidEntry is a trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z260517-1EA | 4061832257754 |