Documenti fondamentali
BAH182130000
SP Bel-Art® Plastic Spring Style Tubing Clamps
for tubing o.d., up to 5/16 in. (8 mm)
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.43
Caratteristiche
for tubing o.d. up to 5/16 in. (8 mm)
Confezionamento
pack of 3 ea
Produttore/marchio commerciale
Bel-Art H18213-0000
Descrizione generale
Perfect for all flexible tubing, this positive-action, self-closing tube clamp opens by squeezing the handles together.
Caratteristiche e vantaggi
  • Chemical and stain-resistant
  • Made of Acetal plastic and stainless steel
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)