BAF371110238
SP Bel-Art® Spinbar® magnetic stir bar
cylindrical white PTFE-coated, L × diam. 2 in. (50.8 mm) × 0.375 in. (9.5 mm)
Sinonimo/i:
Spinbar® magnetic stir bar
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.51
Prodotti consigliati
Materiali
cylindrical white PTFE-coated
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F37111-0238
L × diam.
2 in. (50.8 mm) × 0.375 in. (9.5 mm)
Categorie correlate
Descrizione generale
- Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
- Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
- Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
- Autoclave safe.
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics.
Leakproof, seamless, polished, and contains a permanent Alnico V magnet. Egg-shape bar is designed especially for use in round-bottom vessels.
Caratteristiche e vantaggi
- Small removable pivot ring in the center adds to their versatility
- Pivot ring minimizes the contact area of the bar to the vessel, reduces friction and lessens marring of plastic containers
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
- Autoclave safe
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)