Merck
Z740952
Sigma-Aldrich
Aldrich® Essentials culture tube with screw cap and PTFE/rubber liner
capacity 50 mL, O.D. × L 25 mm × 150 mm, round bottom
About This Item
UNSPSC-Code:
41121706
NACRES:
NB.11
Materialien
borosilicate glass tube
round bottom
screw top cap
screw top neck
Leistungsmerkmale
neck thread external thread
Verpackung
pack of 48 ea
Hersteller/Markenname
Sigma-Aldrich
AD × L
25 mm × 150 mm
Kapazität
50 mL
Allgemeine Beschreibung
Clear borosilicate glass, round bottom culture tubes with thread top and black phenolic cap. Cap has bonded PTFE faced rubber liner.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z740952-48EA4061833641576