Merck
SYNA192424
Synthware connecting adapter with PTFE stopcock
joint: ST/NS 24/40
Synonym(e):
Synthware A192424, Glass connecting adapter
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
joint: ST/NS24/40
Verpackung
pack of 1 ea
Hersteller/Markenname
A192424
Verwandte Kategorien
Allgemeine Beschreibung
Synthware connecting adapter with 2mm stopcock converts standard vessels to airless applications. Standard connecting join (24/40) to fit various laboratory glassware.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)