Documentos Principais
22491
Supelco
Supeltex M-4 Packed Column Ferrule, 1/8 in. Column O.D.
I.D. 1/8 in., 1/8 in. O.D. Column, pkg of 10 ea
About This Item
Código UNSPSC:
41121815
embalagem
pkg of 10 ea
temperatura máxima
450 °C
D.I.
1/8 in.
compatibilidade
1/8 in. O.D. Column
Descrição geral
Ferrules are suitable for the attachment of the column and other glass components to the system. Flexible graphite ferrules withstand high temperatures and are capable of compression.
Supelco has refined the design of these graphite ferrules to provide a clean, sharp profile with minimal flash.
Characteristics:
Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation.
Material Composition: flexible graphite
Maximum Temperature: 450 °C
Informações legais
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Walter Jennings
Gas Chromatography with Glass Capillary Columns (2012)