Documentos Principais
P0614
Sigma-Aldrich
Pipettor Tri-/Clamp-ette
blue
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
fabricante/nome comercial
Heathrow Scientific HS20162A
cor
blue
Descrição geral
Conveniently holds 3 pipettors. An extension to the Clamp-ette family of pipetting aids. This innovative product clamps down securely to table tops or benches and holds most major brands of pipettors. The Tri-Clamp-ette provides the flexibility of housing expensive pipettors practically anywhere in the lab. A screw on each end of the product provides a safe hold with just a few easy turns. Available in blue, red, or purple.
Informações legais
Clamp-ette is a trademark of Heathrow Scientific
Escolha uma das versões mais recentes:
Certificados de análise (COA)