About This Item
Código UNSPSC:
41116107
NACRES:
NB.51
Produtos recomendados
Materiais
stainless steel
Descrição geral
SAFC has qualified EZ BioPac, from ILC Dover as their preferred bulk powder package. Manufactured with Armorflex 114 film, the EzBiopac range is a fit for purpose, scalable and intuitive offer that enables the clean, safe and efficient delivery of bulk powders into bioprocess.
This is an accessory which can be used to assist with the handling of the EZ BioPac by ILC Dover filled with cell culture media.
This is an accessory which can be used to assist with the handling of the EZ BioPac by ILC Dover filled with cell culture media.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
HW11230-01 | 4061829452421 |