GBL664293
Grace Bio-Labs silicone isolator sheet material
thickness 1.6 mm, black silicone sheet, no PSA
Sinônimo(s):
non-adhesive isolator, silicone blocker
About This Item
Código UNSPSC:
41122600
NACRES:
NB.17
Produtos recomendados
Materiais
red silicon
Características
black silicone sheet, no PSA
embalagem
pack of 5
fabricante/nome comercial
Grace Bio-Labs 664293
C × L
13 cm × 18 cm
180 mm × 130 mm
espessura
1.6 mm
Categorias relacionadas
Descrição geral
Silicone isolator sheet material allows researchers to create their own removable hydrophobic barriers to isolate specimens. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended.Silicone sheet material is also available in CultureWell “water-clear” silicone, which is highly recommended for sensitive cell culture applications.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
GBL664293-5EA |