Documentos Principais
EPM1194-9905
Platform for Eppendorf® New Brunswick Innova® 2100/40/40R/42/42R & Excella® E24/E24R Shakers
for 250 mL Erlenmeyer flasks, dedicated, phenolic resin
About This Item
Código UNSPSC:
41103816
NACRES:
NB.73
Características
25 x 250 mL Erlenmeyer flasks
dedicated
for 250 mL Erlenmeyer flasks
phenolic resin
fabricante/nome comercial
Eppendorf® M1194-9905 (Global)
C × L
45.7 cm × 45.7 cm
Categorias relacionadas
Descrição geral
Platforms designed for the Eppendorf New Brunswick Innova 2100/40/40R/42/42R & Excella E24/E24R shakers to ensure a safe and perfect fit.
Informações legais
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc. 
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Escolha uma das versões mais recentes:
Certificados de análise (COA)