Key Documents
EP5702704008
Eppendorf® Fixed-angle rotor
F-35-30-17, 30 x 15 ml, incl. 20 adapters Falcon tubes and 30 rubber mats
About This Item
Código UNSPSC:
41103909
NACRES:
NB.82
Características
to hold 20 x 15 mL conical tubes
to hold 30 x 15 mL glass tubes
F-35-30-17, 30 x 15 ml, incl. 20 adapters Falcon tubes and 30 rubber mats
embalagem
pack of 1 ea
fabricante/nome comercial
Eppendorf® 5702704008
Parâmetros
4,400 max. RCF (2,750 xg)
técnica(s)
centrifugation: suitable
compatibilidade
for use with centrifuge models 5702/5702R/5702RH
Informações legais
Eppendorf is a registered trademark of Eppendorf AG