Key Documents
EP5409715003
Rotor for Eppendorf® 5427R Centrifuge
S-24-11-AT, aerosol-tight, to hold, 24 x 1.5/2.0 mL tubes, with lid and wind shield
About This Item
Código UNSPSC:
41103909
NACRES:
NB.82
Características
to hold 24 x 1.5/2.0 mL tubes
aerosol-tight
swing bucket rotor
with lid and wind shield
embalagem
pack of 1 ea
fabricante/nome comercial
Eppendorf® 5409715003 (Global)
Parâmetros
12,700 rpm
16,050 × g max. RCF
técnica(s)
centrifugation: suitable
compatibilidade
for use with centrifuge models 5427R
Informações legais
Eppendorf is a registered trademark of Eppendorf AG