Key Documents
CLS2975246
Corning® cover glasses
rectangular, No. 1, W × L 24 mm × 60 mm
Sinônimo(s):
coverslips, glass covers, slide covers
About This Item
Código UNSPSC:
41121800
NACRES:
NB.16
Materiais
borosilicate glass
rectangular
embalagem
pack of 100 ea
case of 1000 ea
fabricante/nome comercial
Corning 2975-246
L × C
24 mm × 60 mm
tipo de ligação
non-treated surface
Características e benefícios
  • Corning® cover glasses are made from optically clear borosilicate glass.
  • Chemically resistant cover glasses are not affected by weathering or surface attachment.
Informações legais
Corning is a registered trademark of Corning, Inc.
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 9
1 of 9
Corning® cover glasses square, No. 1, W x L 22 mm x mm
CLS284522
Corning® cover glasses
BRAND® cover glass L × W 24 mm × 50 mm, rectangular
BR470819
BRAND® cover glass
Corning® cover glasses square, No. 2, W × L 25 mm × 25 mm
CLS285525
Corning® cover glasses
BRAND® cover glass L × W 24 mm × 40 mm, rectangular
BR470816
BRAND® cover glass
Corning® cover glasses rectangular, No. 11/2, W × L 22 mm × 50 mm
CLS2980225
Corning® cover glasses
Corning® cover glasses square, No. 11/2, W × L 22 mm × 22 mm
CLS285022
Corning® cover glasses
Cover glasses size 24 mm × 50 mm
C8181
Cover glasses
Cover glasses size 24 mm × 40 mm
C7931
Cover glasses
Cover glasses size 22 mm × 22 mm
C9802
Cover glasses