Documentos Principais
CLS2975224
Corning® cover glasses
rectangular, No. 1, W × L 22 mm × 40 mm
Sinônimo(s):
coverslips, glass covers, slide covers
About This Item
Código UNSPSC:
41121800
NACRES:
NB.16
Materiais
borosilicate glass
rectangular
embalagem
pack of 100 ea
case of 1000 ea
fabricante/nome comercial
Corning 2975-224
L × C
22 mm × 40 mm
tipo de ligação
non-treated surface
Características e benefícios
  • Corning® cover glasses are made from optically clear borosilicate glass.
  • Chemically resistant cover glasses are not affected by weathering or surface attachment.
Informações legais
Corning is a registered trademark of Corning, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 2
1 of 2
BRAND® cover glass L × W 22 mm × 22 mm, square
BR470055
BRAND® cover glass
Fluoroshield™ histology mounting medium
Sigma-Aldrich
F6182
Fluoroshield
Global Trade Item Number
SKUGTIN
CLS2975224-1000EA4061833882344
CLS2975224-100EA4061833882351