Documentos Principais
MZPNNVX00
Millipore
Multiscreen® 384 well Plate
V-bottom clear natural polypropylene plate, non-sterile, pkg of 100 plates
About This Item
Código UNSPSC:
41122102
NACRES:
NB.15
Materiais
V-bottom clear natural polypropylene plate
Nível de qualidade
esterilidade
non-sterile
linha de produto
MultiScreen®
embalagem
pkg of 100 plates
tamanho da placa
384 wells
tamanho
384 well
Descrição geral
MultiScreen® 384-well polypropylene microplates meet industry standard dimensions for use in common biochemical assay platforms, including automated liquid handling, microplate readers, and other microplate compatible devices. Polypropylene microplates are appropriate for sample preparation and storage in addition to some biochemical assays.
forma física
Color Code:Natural
Outras notas
Automation Compatible:Yes
Informações legais
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany