Z680486
Duran® GL 45 connection system caps and accessories
insert for hose connection screw cap GL 14, 3.2 mm
Sinônimo(s):
1129817
About This Item
Código UNSPSC:
41121800
NACRES:
NB.17
Produtos recomendados
Materiais
white PTFE/silicone
fabricante/nome comercial
DURAN® 1129817
Parâmetros
-50-230 °C temp. limit
Categorias relacionadas
Descrição geral
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Sterile pressure equalization is possible through use of membrane filter.
Características e benefícios
- Manufactured from PP, PTFE, silicone
- Accessories designed for use with the DURAN® GL 45 2/3 port tubing connection caps or GL 45 / GLS 80® Stirred Bottle Reactor caps
- Safe and secure liquid transfer into or out of laboratory bottles
- Four inserts are suitable for rigid or semi-rigid tubing with a range of outer diameters: 1.6, 3.0, 3.2 or 6.0 mm
- Open-topped blue PP GL 14 pass-through tubing cap for use with the inserts
- Closed-top red PBT GL 14 cap for sealing unused ports
- Sterile pressure equilisation is possible with pressure compensation set
Informações legais
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z680486-1EA | 4061826711477 |