Key Documents
Z507199
Supelco
Michel-Miller Swagelok® adapter
NPTF 1/8 in., Nylon, Ace-Thred for #25
About This Item
Código UNSPSC:
12000000
NACRES:
SB.52
Materiais
Nylon
fabricante/nome comercial
Ace Glass 584422
técnica(s)
LPLC: suitable
NPTF
1/8 in.
conector
Ace-Thred for #25
Descrição geral
Use to make Swagelok tubing connections with Ace threaded glassware. One end is Ace-Thred, other is NPTF. Connects tubing to M-M columns. Adapters with #50 Ace-Thred fit Ace large columns.
Informações legais
Swagelok is a registered trademark of Swagelok Company
Escolha uma das versões mais recentes:
Certificados de análise (COA)